Recombinat Protein,Rhesus macaque EGF R Protein, His Tag,Rhesus macaque EGF R, His Tag (EGR-C52H1) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P55245).This protein carries a polyhistidine tag at the C-term i 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Rhesus macaque EGF R Protein, His Tag,Rhesus macaque EGF R, His Tag (EGR-C52H1) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P55245).This protein carries a polyhistidine tag at the C-term i 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rhesus macaque EGF R Protein, His Tag,Rhesus macaque EGF R, His Tag (EGR-C52H1) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P55245).This protein carries a polyhistidine tag at the C-term i 1 * 1 mg ACRBEGR-C52H1-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou