Recombinat Protein,Human ENPP-2 Autotaxin Protein, His Tag,Human ENPP-2, His Tag (EN2-H5241) is expressed from human 293 cells (HEK 293). It contains AA Asp 49 - Ile 863 (Accession NP001035181).This protein carries a polyhistidine tag at the N-termi 1 * 250 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human ENPP-2 Autotaxin Protein, His Tag,Human ENPP-2, His Tag (EN2-H5241) is expressed from human 293 cells (HEK 293). It contains AA Asp 49 - Ile 863 (Accession NP001035181).This protein carries a polyhistidine tag at the N-termi 1 * 250 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human ENPP-2 Autotaxin Protein, His Tag,Human ENPP-2, His Tag (EN2-H5241) is expressed from human 293 cells (HEK 293). It contains AA Asp 49 - Ile 863 (Accession NP001035181).This protein carries a polyhistidine tag at the N-termi 1 * 250 µG ACRBEN2-H5241250UG 1 * 250 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou