Recombinat Protein,Human EphB4 Protein, His Tag,Human EphB4, His Tag (EP4-H5229) is expressed from human 293 cells (HEK293). It cont ains AA Leu 16 - Ala 539 (Accession NP004435.3).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human EphB4 Protein, His Tag,Human EphB4, His Tag (EP4-H5229) is expressed from human 293 cells (HEK293). It cont ains AA Leu 16 - Ala 539 (Accession NP004435.3).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg ACRBEP4-H5229-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou