Human EphA4, His Tag (EP4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Thr 547 (Accession P54764-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human EphA4, His Tag (EP4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Thr 547 (Accession P54764-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human EphA4, His Tag (EP4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Thr 547 (Accession P54764-1). 1 * 100 µG ACRBEP4-H52H8100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou