MABSol® Biotinylated Human EphB4, His Tag, primary amine labeling (EP4-H8229) is expressed from human HEK293 cells. It contains AA L eu 16 - Ala 539 (Accession NP004435.3). It is the biotinylated form of Human EphB4, His Tag (EP4-H5229). 1 * 25 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human EphB4, His Tag, primary amine labeling (EP4-H8229) is expressed from human HEK293 cells. It contains AA L eu 16 - Ala 539 (Accession NP004435.3). It is the biotinylated form of Human EphB4, His Tag (EP4-H5229). 1 * 25 µG ACRBEP4-H8229-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou