Recombinat Protein,Rhesus macaque EpCAM TROP1 Protein, His Tag,Rhesus macaque EpCAM, His Tag (EPM-C5227) is expressed from human 29 3 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession Q1WER1).This protein carries a polyhistidine tag at the e 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Rhesus macaque EpCAM TROP1 Protein, His Tag,Rhesus macaque EpCAM, His Tag (EPM-C5227) is expressed from human 29 3 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession Q1WER1).This protein carries a polyhistidine tag at the e 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rhesus macaque EpCAM TROP1 Protein, His Tag,Rhesus macaque EpCAM, His Tag (EPM-C5227) is expressed from human 29 3 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession Q1WER1).This protein carries a polyhistidine tag at the e 1 * 50 µG ACRBEPM-C5227-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou