Recombinat Protein,Human EpCAM TROP1 Protein, His Tag,Human EpCAM, His Tag (EPM-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human EpCAM TROP1 Protein, His Tag,Human EpCAM, His Tag (EPM-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG ACRBEPM-H5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou