Recombinat Protein,Human EpCAM TROP1 Protein, Fc Tag,Human EpCAM, Fc Tag (EPM-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a human IgG1 Fc tag at the C-terminus. The ca 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human EpCAM TROP1 Protein, Fc Tag,Human EpCAM, Fc Tag (EPM-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a human IgG1 Fc tag at the C-terminus. The ca 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human EpCAM TROP1 Protein, Fc Tag,Human EpCAM, Fc Tag (EPM-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a human IgG1 Fc tag at the C-terminus. The ca 1 * 1 mg ACRBEPM-H5254-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou