MABSol® Biotinylated Human EpCAM, His Tag, primary amine labeling (EPM-H8223) is expressed from human HEK293 cells. It contains AA Gln 24 - Lys 265 (Accession AAH14785.1). It is the biotinylated form of Human EpCAM, His Tag (EPM-H5223). null
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human EpCAM, His Tag, primary amine labeling (EPM-H8223) is expressed from human HEK293 cells. It contains AA Gln 24 - Lys 265 (Accession AAH14785.1). It is the biotinylated form of Human EpCAM, His Tag (EPM-H5223). null ACRBEPM-H8223-25UG Each
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou