Recombinat Protein,Biotinylated Human EpCAM TROP1 Protein, Avitag,His Tag,Biotinylated Human EpCAM, Avitag,His Tag (EPM-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human EpCAM TROP1 Protein, Avitag,His Tag,Biotinylated Human EpCAM, Avitag,His Tag (EPM-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human EpCAM TROP1 Protein, Avitag™,His Tag,Biotinylated Human EpCAM, Avitag,His Tag (EPM-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a 1 * 25 µG ACRBEPM-H82E8-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou