Human ErbB3, His Tag (ER3-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human ErbB3, His Tag (ER3-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human ErbB3, His Tag (ER3-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). 1 * 100 µG ACRBER3-H5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou