MABSol Biotinylated Human ErbB3, His Tag, primary amine labeling (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession NP001973.2). It is the biotinylated form of Human ErbB3, His Tag (ER3-H5223). null
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Human ErbB3, His Tag, primary amine labeling (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession NP001973.2). It is the biotinylated form of Human ErbB3, His Tag (ER3-H5223). null
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human ErbB3, His Tag, primary amine labeling (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession NP001973.2). It is the biotinylated form of Human ErbB3, His Tag (ER3-H5223). null ACRBER3-H8223200UG Each
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou