Recombinat Protein,FITC-Labeled Human ErbB3 Her3 Protein, His Tag,FITC-Labeled Human ErbB3, His Tag (ER3-HF2H5) is expressed from h uman 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). It is the FITC labeled form of Human E 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,FITC-Labeled Human ErbB3 Her3 Protein, His Tag,FITC-Labeled Human ErbB3, His Tag (ER3-HF2H5) is expressed from h uman 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). It is the FITC labeled form of Human E 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,FITC-Labeled Human ErbB3 Her3 Protein, His Tag,FITC-Labeled Human ErbB3, His Tag (ER3-HF2H5) is expressed from h uman 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). It is the FITC labeled form of Human E 1 * 25 µG ACRBER3-HF2H5-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou