Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession NP005226.1) . 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession NP005226.1) . 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession NP005226.1) . 1 * 1 mg ACRBER4-H5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou