Recombinat Protein,Human FABP1 L-FABP Protein, His Tag,Human FABP1, His Tag (FA1-H5148) is expressed from E.coli cells. It contains AA Ser 2 - Ile 127 (Accession NP001434).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human FABP1 L-FABP Protein, His Tag,Human FABP1, His Tag (FA1-H5148) is expressed from E.coli cells. It contains AA Ser 2 - Ile 127 (Accession NP001434).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg ACRBFA1-H5148-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou