Recombinat Protein,Human FABP2 I-FABP Protein, His Tag,Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession NP000125).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human FABP2 I-FABP Protein, His Tag,Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession NP000125).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 200 µG ACRBFA2-H5149200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou