Recombinat Protein,Human FABP5 E-FABP Protein, His Tag,Human FABP5, His Tag (FA5-H5140) is expressed from E.coli cells. It contains AA Ala 2 - Glu 135 (Accession AAH19385).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human FABP5 E-FABP Protein, His Tag,Human FABP5, His Tag (FA5-H5140) is expressed from E.coli cells. It contains AA Ala 2 - Glu 135 (Accession AAH19385).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 100 µG ACRBFA5-H5140100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou