Human Fas Ligand, His Tag (active trimer) (MALS verified) (FAL-H5241) is expressed from human 293 cells (HEK293). It contains AA Pro 134 - Leu 281 (Accession AAH17502.1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Fas Ligand, His Tag (active trimer) (MALS verified) (FAL-H5241) is expressed from human 293 cells (HEK293). It contains AA Pro 134 - Leu 281 (Accession AAH17502.1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Fas Ligand, His Tag (active trimer) (MALS verified) (FAL-H5241) is expressed from human 293 cells (HEK293). It contains AA Pro 134 - Leu 281 (Accession AAH17502.1). 1 * 1 mg ACRBFAL-H5241-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou