Recombinat Protein,Human Fas TNFRSF6 CD95 Protein, His Tag,Human Fas, His Tag (FAS-H5229) is expressed from human 293 cells (HEK29 3). It contains AA Gln 26 - Asn 173 (Accession AAH12479.1).This protein carries a polyhistidine tag at the C-terminus. 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Fas TNFRSF6 CD95 Protein, His Tag,Human Fas, His Tag (FAS-H5229) is expressed from human 293 cells (HEK29 3). It contains AA Gln 26 - Asn 173 (Accession AAH12479.1).This protein carries a polyhistidine tag at the C-terminus. 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Fas TNFRSF6 CD95 Protein, His Tag,Human Fas, His Tag (FAS-H5229) is expressed from human 293 cells (HEK29 3). It contains AA Gln 26 - Asn 173 (Accession AAH12479.1).This protein carries a polyhistidine tag at the C-terminus. 1 * 50 µG ACRBFAS-H5229-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou