Biotinylated Mouse Fc gamma RIV, His,Avitag (BLI verified) (FC4-M82E8) is expressed from human 293 cells (HEK293). It contains AA Gl y 21 - Gln 203 (Accession A0A0B4J1G0-1). 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Mouse Fc gamma RIV, His,Avitag (BLI verified) (FC4-M82E8) is expressed from human 293 cells (HEK293). It contains AA Gl y 21 - Gln 203 (Accession A0A0B4J1G0-1). 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Mouse Fc gamma RIV, His,Avitag (BLI verified) (FC4-M82E8) is expressed from human 293 cells (HEK293). It contains AA Gl y 21 - Gln 203 (Accession A0A0B4J1G0-1). 1 * 200 µG ACRBFC4-M82E8200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou