Biotinylated Human CD64, His,Avitag (SPR BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human CD64, His,Avitag (SPR BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human CD64, His,Avitag (SPR  BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 25 µG ACRBFCA-H82E8-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou