Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It cont ains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region 97 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It cont ains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region 97 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It cont ains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region 97 1 * 50 µG ACRBFCM-C52W9-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou