Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified), is expressed from human 293 cells (HEK2 93). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In a 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified), is expressed from human 293 cells (HEK2 93). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In a 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified), is expressed from human 293 cells (HEK2 93). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In a 1 * 200 µG ACRBFCM-C82W3200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou