MABSol Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,AvitagStrep II Tag (BLIverified) (FCM-C82W5) is expre ssed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FC T 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,AvitagStrep II Tag (BLIverified) (FCM-C82W5) is expre ssed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FC T 1 * 25 G
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,AvitagStrep II Tag (BLI verified) (FCM-C82W5) is expre ssed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FC T 1 * 25 µG ACRBFCM-C82W5-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou