Biotinylated Human FCGRTB2M, His TagStrep II Tag, primary amine labeling (FCM-H8286) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession P55899-1 (FCGRT) P61769-1 (B2M)). It is the orm 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human FCGRTB2M, His TagStrep II Tag, primary amine labeling (FCM-H8286) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession P55899-1 (FCGRT) P61769-1 (B2M)). It is the orm 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human FCGRTB2M, His TagStrep II Tag, primary amine labeling (FCM-H8286) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession P55899-1 (FCGRT) P61769-1 (B2M)). It is the orm 1 * 25 µG ACRBFCM-H8286-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou