MABSol Biotinylated Rat FCGRTB2M Heterodimer Protein, Avitag,His TagStrep II Tag (SPR verified) (FCM-R82W7) is expressed from human HEK293 cells. It contains AA Ala 23 - Ser 298 (FCGRT) Ile 21 - Met 119 (B2M) (Accession P13599-1 (FCGRT) P07151-1 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Rat FCGRTB2M Heterodimer Protein, Avitag,His TagStrep II Tag (SPR verified) (FCM-R82W7) is expressed from human HEK293 cells. It contains AA Ala 23 - Ser 298 (FCGRT) Ile 21 - Met 119 (B2M) (Accession P13599-1 (FCGRT) P07151-1 1 * 200 G
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Rat FCGRTB2M Heterodimer Protein, Avitag,His TagStrep II Tag (SPR verified) (FCM-R82W7) is expressed from human HEK293 cells. It contains AA Ala 23 - Ser 298 (FCGRT) Ile 21 - Met 119 (B2M) (Accession P13599-1 (FCGRT) P07151-1 1 * 200 µG ACRBFCM-R82W7200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou