Recombinat Protein,Human Fetuin B FETUA Protein, His Tag,Human Fetuin B, His Tag (FEB-H52H7) is expressed from human 293 cells (HEK 293). It contains AA Cys 16 - Pro 382 (Accession AAH69670 ).This protein carries a polyhistidine tag at the C-termin n 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Fetuin B FETUA Protein, His Tag,Human Fetuin B, His Tag (FEB-H52H7) is expressed from human 293 cells (HEK 293). It contains AA Cys 16 - Pro 382 (Accession AAH69670 ).This protein carries a polyhistidine tag at the C-termin n 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Fetuin B FETUA Protein, His Tag,Human Fetuin B, His Tag (FEB-H52H7) is expressed from human 293 cells (HEK 293). It contains AA Cys 16 - Pro 382 (Accession AAH69670 ).This protein carries a polyhistidine tag at the C-termin n 1 * 100 µG ACRBFEB-H52H7100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou