Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 1 mg ACRBFG1-H52HY-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou