Recombinat Protein,Biotinylated Human FGL1 Protein, His,Avitag (recommended for biopanning),Biotinylated Human FGL1, His,Avitag (re commended for biopanning) (FG1-H82Ey) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Ac 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human FGL1 Protein, His,Avitag (recommended for biopanning),Biotinylated Human FGL1, His,Avitag (re commended for biopanning) (FG1-H82Ey) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Ac 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human FGL1 Protein, His,Avitag™ (recommended for biopanning),Biotinylated Human FGL1, His,Avitag (re commended for biopanning) (FG1-H82Ey) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Ac 1 * 25 µG ACRBFG1-H82EY-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou