Recombinat Protein,Human FGF-9 Protein, Fc Tag,Recombinant human FGF9 Fc chimera (rhFGF9-Fc) Leu4-Ser208 (Accession AAI03979) was p roduced in human HEK293 cells at ACRObiosystems.This protein carries a human IgG1 Fc tag at the N-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human FGF-9 Protein, Fc Tag,Recombinant human FGF9 Fc chimera (rhFGF9-Fc) Leu4-Ser208 (Accession AAI03979) was p roduced in human HEK293 cells at ACRObiosystems.This protein carries a human IgG1 Fc tag at the N-terminus. The protei 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human FGF-9 Protein, Fc Tag,Recombinant human FGF9 Fc chimera (rhFGF9-Fc) Leu4-Ser208 (Accession AAI03979) was p roduced in human HEK293 cells at ACRObiosystems.This protein carries a human IgG1 Fc tag at the N-terminus. The protei 1 * 1 mg ACRBFG9-H4267-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou