Recombinat Protein,Human FKBP4 FKBP52 Protein, His,PA Tag,Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HE K293). It contains AA Met 1 - Ala 459 (Accession AAH01786).This protein carries a polyhistidine tag at the C-terminu 1 * 500 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human FKBP4 FKBP52 Protein, His,PA Tag,Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HE K293). It contains AA Met 1 - Ala 459 (Accession AAH01786).This protein carries a polyhistidine tag at the C-terminu 1 * 500 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human FKBP4 FKBP52 Protein, His,PA Tag,Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HE K293). It contains AA Met 1 - Ala 459 (Accession AAH01786).This protein carries a polyhistidine tag at the C-terminu 1 * 500 µG ACRBFK4-H52Q3500UG 1 * 500 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou