Recombinat Protein,Human FKBP12 FKBP1A Protein, His Tag,Human FKBP12, His Tag (FKA-H5122) is expressed from E.coli cells. It contai ns AA Met 1 - Glu 108 (Accession AAH05147).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human FKBP12 FKBP1A Protein, His Tag,Human FKBP12, His Tag (FKA-H5122) is expressed from E.coli cells. It contai ns AA Met 1 - Glu 108 (Accession AAH05147).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human FKBP12 FKBP1A Protein, His Tag,Human FKBP12, His Tag (FKA-H5122) is expressed from E.coli cells. It contai ns AA Met 1 - Glu 108 (Accession AAH05147).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg ACRBFKA-H5122-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou