Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acc ession P35916-1). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acc ession P35916-1). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acc ession P35916-1). 1 * 25 µG ACRBFL4-H82E1-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou