Human Flt-3 Ligand, His Tag (FLL-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Accession P497 71-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Flt-3 Ligand, His Tag (FLL-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Accession P497 71-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Flt-3 Ligand, His Tag (FLL-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Accession P497 71-1). 1 * 1 mg ACRBFLL-H5223-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou