Cynomolgus Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (A ccession NP001181576.1). In the region Arg 25 - Met 233, the AA sequence of Cynomolgus and Rhesus macaque FOLR1 are 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (A ccession NP001181576.1). In the region Arg 25 - Met 233, the AA sequence of Cynomolgus and Rhesus macaque FOLR1 are 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (A ccession NP001181576.1). In the region Arg 25 - Met 233, the AA sequence of Cynomolgus and Rhesus macaque FOLR1 are 1 * 100 µG ACRBFO1-C52H8100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou