Recombinat Protein,Human FOLR1 Protein, Strep Tag,Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession NP057937.1).This protein carries a twin strep tag at the C-terminus. The prot 1 * 250 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human FOLR1 Protein, Strep Tag,Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession NP057937.1).This protein carries a twin strep tag at the C-terminus. The prot 1 * 250 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human FOLR1 Protein, Strep Tag,Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession NP057937.1).This protein carries a twin strep tag at the C-terminus. The prot 1 * 250 µG ACRBFO1-H528B250UG 1 * 250 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou