Mouse FOLR1, His Tag (FO1-M5225) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession P35846-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse FOLR1, His Tag (FO1-M5225) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession P35846-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse FOLR1, His Tag (FO1-M5225) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession P35846-1). 1 * 1 mg ACRBFO1-M5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou