Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 1 mg ACRBFO2-H5223-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou