Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 100 µG ACRBFO2-H5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou