Recombinat Protein,Human Follistatin-like 1 FSTL1 Protein, His Tag,Human Follistatin-like 1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession AAH00055).This protein carries a polyhistidine t e 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Follistatin-like 1 FSTL1 Protein, His Tag,Human Follistatin-like 1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession AAH00055).This protein carries a polyhistidine t e 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Follistatin-like 1 FSTL1 Protein, His Tag,Human Follistatin-like 1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession AAH00055).This protein carries a polyhistidine t e 1 * 1 mg ACRBFS1-H5227-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou