Recombinat Protein,Human Follistatin-like 1 FSTL1 Protein, His Tag,Human Follistatin-like 1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession AAH00055).This protein carries a polyhistidine t e 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Follistatin-like 1 FSTL1 Protein, His Tag,Human Follistatin-like 1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession AAH00055).This protein carries a polyhistidine t e 1 * 100 µG ACRBFS1-H5227100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou