Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HE K293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HE K293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HE K293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 µG ACRBFZ5-H5259100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou