Recombinat Protein,Human Frizzled-7 FZD7 Protein, Fc Tag,Human Frizzled-7, Fc Tag (FZ7-H5258) is expressed from human 293 cells (HE K293). It contains AA Gln 33 - Leu 185 (Accession AAH15915).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Frizzled-7 FZD7 Protein, Fc Tag,Human Frizzled-7, Fc Tag (FZ7-H5258) is expressed from human 293 cells (HE K293). It contains AA Gln 33 - Leu 185 (Accession AAH15915).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Frizzled-7 FZD7 Protein, Fc Tag,Human Frizzled-7, Fc Tag (FZ7-H5258) is expressed from human 293 cells (HE K293). It contains AA Gln 33 - Leu 185 (Accession AAH15915).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 µG ACRBFZ7-H5258100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou