Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termi 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termi 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG ACRBFZ7-H52H9100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou