Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contai ns AA Met 1 - Ile 250 (Accession NP002297).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contai ns AA Met 1 - Ile 250 (Accession NP002297).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 200 µG ACRBGA3-H5129200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou