Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5247) is expressed from human 293 cells (HEK293 ). It contains AA Met 1 - Ile 250 (Accession P17931-1).This protein carries a polyhistidine tag at the N-terminus. T 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5247) is expressed from human 293 cells (HEK293 ). It contains AA Met 1 - Ile 250 (Accession P17931-1).This protein carries a polyhistidine tag at the N-terminus. T 1 * 1 mg ACRBGA3-H5247-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou