Recombinat Protein,Human GAPDH G3PDH Protein, His Tag,Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession AAH01601).This protein carries a polyhistidine tag at the N-terminus. The protein has a 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human GAPDH G3PDH Protein, His Tag,Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession AAH01601).This protein carries a polyhistidine tag at the N-terminus. The protein has a 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human GAPDH G3PDH Protein, His Tag,Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession AAH01601).This protein carries a polyhistidine tag at the N-terminus. The protein has a 1 * 1 mg ACRBGAH-H5145-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou