Recombinat Protein,Human GFR alpha-1 Protein, His Tag,Human GFR alpha-1, His Tag (GF1-H5224) is expressed from human 293 cells (HEK2 93). It contains AA Asp 25 - Ser 424 (Accession NP665736.1).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human GFR alpha-1 Protein, His Tag,Human GFR alpha-1, His Tag (GF1-H5224) is expressed from human 293 cells (HEK2 93). It contains AA Asp 25 - Ser 424 (Accession NP665736.1).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human GFR alpha-1 Protein, His Tag,Human GFR alpha-1, His Tag (GF1-H5224) is expressed from human 293 cells (HEK2 93). It contains AA Asp 25 - Ser 424 (Accession NP665736.1).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg ACRBGF1-H5224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou