Recombinat Protein,Human GFR alpha-like Protein, His Tag,Human GFR alpha-like, His Tag (GFA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Glu 351 (Accession Q6UXV0-1).This protein carries a polyhistidine tag at the C-te t 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human GFR alpha-like Protein, His Tag,Human GFR alpha-like, His Tag (GFA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Glu 351 (Accession Q6UXV0-1).This protein carries a polyhistidine tag at the C-te t 1 * 100 µG ACRBGFA-H52H3100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou