MABSol Biotinylated Human GHR, His Tag, ultra sensitivity (primary amine labeling) (GHR-H8222) is expressed from human HEK293 cells . It contains AA Ala 27 - Tyr 264 (Accession P10912-1). It is the biotinylated form of Human Growth Hormone R (GHR) 2 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Human GHR, His Tag, ultra sensitivity (primary amine labeling) (GHR-H8222) is expressed from human HEK293 cells . It contains AA Ala 27 - Tyr 264 (Accession P10912-1). It is the biotinylated form of Human Growth Hormone R (GHR) 2 1 * 50 G
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human GHR, His Tag, ultra sensitivity (primary amine labeling) (GHR-H8222) is expressed from human HEK293 cells . It contains AA Ala 27 - Tyr 264 (Accession P10912-1). It is the biotinylated form of Human Growth Hormone R (GHR) 2 1 * 50 µG ACRBGHR-H8222-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou